Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OBSL1
Entrez Gene ID
23363
Gene Name
obscurin like 1
Gene Aliases
-
Chromosome Location
Chr.2: 219549408 - 219571573 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015311.2 NP_056126.1 15 - 16 5192 79
XM_005246424.4 XP_005246481.1 14 - 15 4940 79
XM_011510857.2 XP_011509159.1 15 - 16 5219 79
XM_017003696.1 XP_016859185.1 15 - 16 5218 79
XM_017003697.1 XP_016859186.1 15 - 16 5218 79
XM_017003698.1 XP_016859187.1 14 - 15 4941 79
XM_017003699.1 XP_016859188.1 14 - 15 4942 79
XM_017003700.1 XP_016859189.1 13 - 14 4664 79
GenBank mRNA EF063638.1 - 15 - 16 4935 79

View Full Product Details