Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OBSL1
Entrez Gene ID
23363
Gene Name
obscurin like 1
Gene Aliases
-
Chromosome Location
Chr.2: 219549408 - 219571573 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173431.1 NP_001166902.1 9 - 10 3537 60
NM_015311.2 NP_056126.1 9 - 10 3537 60
XM_005246424.4 XP_005246481.1 9 - 10 3561 60
XM_011510857.2 XP_011509159.1 9 - 10 3564 60
XM_011510863.2 XP_011509165.1 9 - 10 3556 60
XM_011510864.2 XP_011509166.1 9 - 10 3556 60
XM_011510865.2 XP_011509167.1 9 - 10 3556 60
XM_017003696.1 XP_016859185.1 9 - 10 3563 60
XM_017003697.1 XP_016859186.1 9 - 10 3563 60
XM_017003698.1 XP_016859187.1 9 - 10 3562 60
GenBank mRNA AK023854.1 - 1 - 2 204 60
EF063637.1 - 9 - 10 3283 60
EF063638.1 - 9 - 10 3280 60

View Full Product Details