Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC18
Entrez Gene ID
343099
Gene Name
coiled-coil domain containing 18
Gene Aliases
NY-SAR-41
Chromosome Location
Chr.1: 93179802 - 93279037 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306076.1 NP_001293005.1 22 - 23 3245 98
NM_206886.4 NP_996769.3 22 - 23 3248 98
XM_011541361.2 XP_011539663.2 22 - 23 3457 98
XM_017001154.1 XP_016856643.1 22 - 23 3457 98
XM_017001155.1 XP_016856644.1 22 - 23 3454 98
XM_017001156.1 XP_016856645.1 22 - 23 3427 98
XM_017001157.1 XP_016856646.1 22 - 23 3409 98
XM_017001158.1 XP_016856647.1 22 - 23 3454 98
XM_017001159.1 XP_016856648.1 21 - 22 3325 98
XM_017001160.1 XP_016856649.1 22 - 23 3457 98
XM_017001161.1 XP_016856650.1 22 - 23 3457 98
XM_017001162.1 XP_016856651.1 22 - 23 3170 98
XM_017001163.1 XP_016856652.1 22 - 23 3724 98
XM_017001164.1 XP_016856653.1 22 - 23 3268 98
XM_017001165.1 XP_016856654.1 22 - 23 3457 98
XM_017001166.1 XP_016856655.1 20 - 21 2912 98
XM_017001167.1 XP_016856656.1 20 - 21 2847 98
XM_017001168.1 XP_016856657.1 18 - 19 2612 98
XM_017001170.1 XP_016856659.1 9 - 10 2841 98
XM_017001171.1 XP_016856660.1 9 - 10 1139 98
GenBank mRNA AK126045.1 - 16 - 17 2316 98
BX538286.1 - 14 - 15 98

View Full Product Details