Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CCDC18
- Entrez Gene ID
-
343099
- Gene Name
- coiled-coil domain containing 18
- Gene Aliases
-
NY-SAR-41
- Chromosome Location
-
Chr.1: 93179802 - 93279037 on Build GRCh38
View Full Product Details