Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC18
Entrez Gene ID
343099
Gene Name
coiled-coil domain containing 18
Gene Aliases
NY-SAR-41
Chromosome Location
Chr.1: 93179802 - 93279037 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306076.1 NP_001293005.1 2 - 3 300 122
NM_206886.4 NP_996769.3 2 - 3 300 122
XM_011541361.2 XP_011539663.2 2 - 3 509 122
XM_011541372.2 XP_011539674.2 2 - 3 509 122
XM_017001154.1 XP_016856643.1 2 - 3 509 122
XM_017001155.1 XP_016856644.1 2 - 3 509 122
XM_017001156.1 XP_016856645.1 2 - 3 479 122
XM_017001157.1 XP_016856646.1 2 - 3 509 122
XM_017001158.1 XP_016856647.1 2 - 3 509 122
XM_017001159.1 XP_016856648.1 2 - 3 509 122
XM_017001160.1 XP_016856649.1 2 - 3 509 122
XM_017001161.1 XP_016856650.1 2 - 3 509 122
XM_017001162.1 XP_016856651.1 2 - 3 222 122
XM_017001163.1 XP_016856652.1 2 - 3 776 122
XM_017001164.1 XP_016856653.1 2 - 3 320 122
XM_017001165.1 XP_016856654.1 2 - 3 509 122
XM_017001169.1 XP_016856658.1 2 - 3 509 122
GenBank mRNA BC056667.1 - 2 - 3 296 122
BX099893.1 - 2 - 3 270 122

View Full Product Details