Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC18
Entrez Gene ID
343099
Gene Name
coiled-coil domain containing 18
Gene Aliases
NY-SAR-41
Chromosome Location
Chr.1: 93179802 - 93279037 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306076.1 NP_001293005.1 6 - 7 871 91
NM_206886.4 NP_996769.3 6 - 7 871 91
XM_011541361.2 XP_011539663.2 6 - 7 1080 91
XM_011541372.2 XP_011539674.2 6 - 7 1080 91
XM_017001154.1 XP_016856643.1 6 - 7 1080 91
XM_017001155.1 XP_016856644.1 6 - 7 1080 91
XM_017001156.1 XP_016856645.1 6 - 7 1050 91
XM_017001157.1 XP_016856646.1 6 - 7 1080 91
XM_017001158.1 XP_016856647.1 6 - 7 1080 91
XM_017001159.1 XP_016856648.1 6 - 7 1080 91
XM_017001160.1 XP_016856649.1 6 - 7 1080 91
XM_017001161.1 XP_016856650.1 6 - 7 1080 91
XM_017001162.1 XP_016856651.1 6 - 7 793 91
XM_017001163.1 XP_016856652.1 6 - 7 1347 91
XM_017001164.1 XP_016856653.1 6 - 7 891 91
XM_017001165.1 XP_016856654.1 6 - 7 1080 91
XM_017001166.1 XP_016856655.1 4 - 5 535 91
XM_017001167.1 XP_016856656.1 4 - 5 470 91
XM_017001168.1 XP_016856657.1 3 - 4 357 91
XM_017001169.1 XP_016856658.1 6 - 7 1080 91
GenBank mRNA BC056667.1 - 6 - 7 867 91

View Full Product Details