The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304533.1 | NP_001291462.1 | 3 - 4 | 410 | 106 |
NR_130764.1 | 3 - 4 | 410 | 106 | ||
XM_011517511.2 | XP_011515813.1 | 3 - 4 | 410 | 106 | |
XM_011517512.2 | XP_011515814.1 | 3 - 4 | 410 | 106 | |
XM_017013359.1 | XP_016868848.1 | 3 - 4 | 410 | 106 | |
XM_017013360.1 | XP_016868849.1 | 3 - 4 | 410 | 106 | |
XM_017013361.1 | XP_016868850.1 | 3 - 4 | 1792 | 106 | |
GenBank mRNA | AK096949.1 | - | 3 - 4 | 410 | 106 |
AK299689.1 | - | 3 - 4 | 683 | 106 |