The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000049.2 | NP_000040.1 | 1 - 2 | 389 | 83 |
NM_001128085.1 | NP_001121557.1 | 2 - 3 | 322 | 83 | |
XM_017024661.1 | XP_016880150.1 | 3 - 4 | 538 | 83 | |
GenBank mRNA | AK312901.1 | - | 2 - 3 | 380 | 83 |
BC029128.1 | - | 1 - 2 | 366 | 83 | |
CR541908.1 | - | 1 - 2 | 231 | 83 | |
S67156.1 | - | 1 - 2 | 389 | 83 |