The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139072.3 | NP_620711.3 | 10 - 11 | 1871 | 104 |
XM_005246950.2 | XP_005247007.1 | 9 - 10 | 1735 | 104 | |
GenBank mRNA | AF442487.1 | - | 10 - 11 | 1784 | 104 |
AK289553.1 | - | 10 - 11 | 1829 | 104 | |
AK304596.1 | - | 8 - 9 | 1221 | 104 | |
AK304631.1 | - | 9 - 10 | 1223 | 104 | |
AK308576.1 | - | 6 - 7 | 574 | 104 | |
AL137311.1 | - | 3 - 4 | 269 | 104 | |
AY358891.1 | - | 10 - 11 | 1834 | 104 | |
BC024766.2 | - | 6 - 7 | 877 | 104 | |
BC035009.1 | - | 10 - 11 | 1861 | 104 |