The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139072.3 | NP_620711.3 | 5 - 6 | 1143 | 68 |
XM_005246950.2 | XP_005247007.1 | 5 - 6 | 1130 | 68 | |
GenBank mRNA | AF442487.1 | - | 5 - 6 | 1056 | 68 |
AK289553.1 | - | 5 - 6 | 1101 | 68 | |
AK304596.1 | - | 3 - 4 | 68 | ||
AK304631.1 | - | 3 - 4 | 522 | 68 | |
AY358891.1 | - | 5 - 6 | 1106 | 68 | |
BC024766.2 | - | 1 - 2 | 149 | 68 | |
BC035009.1 | - | 5 - 6 | 1133 | 68 |