The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005686.2 | NP_005677.2 | 1 - 2 | 594 | 101 |
XM_005245623.2 | XP_005245680.1 | 1 - 2 | 616 | 101 | |
GenBank mRNA | AF116571.1 | - | 1 - 2 | 107 | 101 |
AK314197.1 | - | 1 - 2 | 90 | 101 | |
BC106038.1 | - | 1 - 2 | 613 | 101 | |
BQ935858.1 | - | 1 - 2 | 585 | 101 |