The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168370.1 | NP_001161842.1 | 15 - 16 | 3376 | 134 |
NM_014780.4 | NP_055595.2 | 15 - 16 | 3369 | 134 | |
XM_005249503.2 | XP_005249560.1 | 15 - 16 | 3232 | 134 | |
XM_006715285.1 | XP_006715348.1 | 15 - 16 | 3470 | 134 | |
XM_011515019.2 | XP_011513321.1 | 15 - 16 | 3327 | 134 | |
XM_011515020.2 | XP_011513322.1 | 15 - 16 | 3230 | 134 | |
XM_011515021.1 | XP_011513323.1 | 10 - 11 | 1411 | 134 | |
XM_017011533.1 | XP_016867022.1 | 15 - 16 | 3355 | 134 | |
XM_017011534.1 | XP_016867023.1 | 15 - 16 | 3354 | 134 | |
XM_017011535.1 | XP_016867024.1 | 15 - 16 | 3258 | 134 | |
XM_017011536.1 | XP_016867025.1 | 15 - 16 | 3497 | 134 | |
XM_017011537.1 | XP_016867026.1 | 15 - 16 | 3470 | 134 | |
XM_017011538.1 | XP_016867027.1 | 15 - 16 | 3401 | 134 | |
XM_017011539.1 | XP_016867028.1 | 15 - 16 | 3374 | 134 | |
GenBank mRNA | AK292806.1 | - | 15 - 16 | 3306 | 134 |
AK302668.1 | - | 15 - 16 | 3313 | 134 | |
AK307936.1 | - | 5 - 6 | 797 | 134 | |
BC033647.1 | - | 15 - 16 | 3351 | 134 | |
D38548.1 | - | 15 - 16 | 3123 | 134 |