The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168370.1 | NP_001161842.1 | 21 - 22 | 4362 | 95 |
NM_014780.4 | NP_055595.2 | 21 - 22 | 4355 | 95 | |
XM_005249503.2 | XP_005249560.1 | 21 - 22 | 4218 | 95 | |
XM_006715285.1 | XP_006715348.1 | 21 - 22 | 4456 | 95 | |
XM_011515019.2 | XP_011513321.1 | 21 - 22 | 4313 | 95 | |
XM_011515020.2 | XP_011513322.1 | 21 - 22 | 4216 | 95 | |
XM_011515021.1 | XP_011513323.1 | 16 - 17 | 2397 | 95 | |
XM_017011533.1 | XP_016867022.1 | 21 - 22 | 4341 | 95 | |
XM_017011534.1 | XP_016867023.1 | 21 - 22 | 4340 | 95 | |
XM_017011535.1 | XP_016867024.1 | 21 - 22 | 4244 | 95 | |
XM_017011536.1 | XP_016867025.1 | 21 - 22 | 4483 | 95 | |
XM_017011537.1 | XP_016867026.1 | 21 - 22 | 4456 | 95 | |
XM_017011538.1 | XP_016867027.1 | 21 - 22 | 4387 | 95 | |
XM_017011539.1 | XP_016867028.1 | 21 - 22 | 4360 | 95 | |
GenBank mRNA | AK292806.1 | - | 21 - 22 | 4292 | 95 |
AK302668.1 | - | 21 - 22 | 4299 | 95 | |
AK307936.1 | - | 11 - 12 | 1783 | 95 | |
BC033647.1 | - | 21 - 22 | 4337 | 95 | |
D38548.1 | - | 21 - 22 | 4109 | 95 |