Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CUL7
Entrez Gene ID
9820
Gene Name
cullin 7
Gene Aliases
3M1, KIAA0076, dJ20C7.5
Chromosome Location
Chr.6: 43037617 - 43053950 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168370.1 NP_001161842.1 9 - 10 2508 57
NM_014780.4 NP_055595.2 9 - 10 2501 57
XM_005249503.2 XP_005249560.1 9 - 10 2364 57
XM_006715285.1 XP_006715348.1 9 - 10 2602 57
XM_011515019.2 XP_011513321.1 9 - 10 2459 57
XM_011515020.2 XP_011513322.1 9 - 10 2362 57
XM_017011533.1 XP_016867022.1 9 - 10 2487 57
XM_017011534.1 XP_016867023.1 9 - 10 2486 57
XM_017011535.1 XP_016867024.1 9 - 10 2390 57
XM_017011536.1 XP_016867025.1 9 - 10 2629 57
XM_017011537.1 XP_016867026.1 9 - 10 2602 57
XM_017011538.1 XP_016867027.1 9 - 10 2533 57
XM_017011539.1 XP_016867028.1 9 - 10 2506 57
XM_017011540.1 XP_016867029.1 9 - 10 2492 57
GenBank mRNA AK292806.1 - 9 - 10 2438 57
AK302668.1 - 9 - 10 2445 57
BC033647.1 - 9 - 10 2483 57
D38548.1 - 9 - 10 2255 57

View Full Product Details