The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020750.2 | NP_065801.1 | 11 - 12 | 1435 | 111 |
GenBank mRNA | AB033117.2 | - | 11 - 12 | 1374 | 111 |
AF271159.1 | - | 11 - 12 | 1224 | 111 | |
AF298880.1 | - | 11 - 12 | 1287 | 111 | |
AK056619.1 | - | 7 - 8 | 760 | 111 | |
AK291452.1 | - | 11 - 12 | 1398 | 111 | |
AL834207.1 | - | 11 - 12 | 1344 | 111 | |
BC009969.2 | - | 4 - 5 | 399 | 111 | |
BC062635.1 | - | 11 - 12 | 1401 | 111 |