The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286138.1 | NP_001273067.1 | 3 - 4 | 649 | 122 |
NM_006714.4 | NP_006705.1 | 4 - 5 | 863 | 122 | |
XM_005266797.4 | XP_005266854.1 | 4 - 5 | 570 | 122 | |
XM_005266799.2 | XP_005266856.1 | 4 - 5 | 863 | 122 | |
GenBank mRNA | AK000184.1 | - | 4 - 5 | 636 | 122 |
AK301365.1 | - | 3 - 4 | 372 | 122 | |
BC018999.2 | - | 4 - 5 | 633 | 122 |