The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286138.1 | NP_001273067.1 | 6 - 7 | 1131 | 102 |
NM_006714.4 | NP_006705.1 | 7 - 8 | 1345 | 102 | |
XM_005266797.4 | XP_005266854.1 | 7 - 8 | 1052 | 102 | |
XM_005266798.2 | XP_005266855.1 | 5 - 6 | 1078 | 102 | |
XM_005266801.2 | XP_005266858.1 | 5 - 6 | 795 | 102 | |
XM_005266802.2 | XP_005266859.1 | 4 - 5 | 2733 | 102 | |
XM_017010204.1 | XP_016865693.1 | 6 - 7 | 947 | 102 | |
GenBank mRNA | AK000184.1 | - | 7 - 8 | 1118 | 102 |
AK301365.1 | - | 6 - 7 | 854 | 102 | |
BC018999.2 | - | 7 - 8 | 1115 | 102 | |
Y08136.1 | - | 2 - 3 | 221 | 102 |