Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHH1
Entrez Gene ID
57475
Gene Name
pleckstrin homology, MyTH4 and FERM domain containing H1
Gene Aliases
-
Chromosome Location
Chr.14: 67533155 - 67589766 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020715.2 NP_065766.1 26 - 27 3837 66
XM_011537009.1 XP_011535311.1 26 - 27 4013 66
XM_011537015.2 XP_011535317.1 19 - 20 4068 66
XM_017021499.1 XP_016876988.1 25 - 26 3994 66
XM_017021500.1 XP_016876989.1 26 - 27 3929 66
XM_017021501.1 XP_016876990.1 25 - 26 3910 66
XM_017021502.1 XP_016876991.1 19 - 20 3790 66
XM_017021503.1 XP_016876992.1 19 - 20 3150 66
XM_017021504.1 XP_016876993.1 21 - 22 3292 66
GenBank mRNA AB033026.1 - 26 - 27 3813 66
AK124787.1 - 3 - 4 894 66
AK293950.1 - 7 - 8 878 66
AK295896.1 - 19 - 20 2537 66
BC014159.2 - 3 - 4 296 66
BC146787.1 - 26 - 27 3813 66
BX648137.1 - 4 - 5 2194 66
BX648464.1 - 17 - 18 2949 66

View Full Product Details