The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271999.1 | NP_001258928.1 | 47 - 48 | 6286 | 136 |
NM_001272000.1 | NP_001258929.1 | 47 - 48 | 6199 | 136 | |
NM_001272001.1 | NP_001258930.1 | 46 - 47 | 6193 | 136 | |
NM_033407.3 | NP_212132.2 | 47 - 48 | 6226 | 136 | |
XM_005271292.2 | XP_005271349.1 | 48 - 49 | 6282 | 136 | |
XM_011542326.2 | XP_011540628.1 | 48 - 49 | 6309 | 136 | |
XM_011542327.2 | XP_011540629.1 | 47 - 48 | 6303 | 136 | |
XM_011542328.2 | XP_011540630.1 | 47 - 48 | 6294 | 136 | |
XM_017002639.1 | XP_016858128.1 | 46 - 47 | 6210 | 136 | |
GenBank mRNA | AB051558.1 | - | 28 - 29 | 3702 | 136 |
AK055493.1 | - | 14 - 15 | 1843 | 136 | |
AK055905.1 | - | 19 - 20 | 2718 | 136 | |
AK091479.1 | - | 21 - 22 | 2693 | 136 | |
AK125049.1 | - | 33 - 34 | 4371 | 136 | |
AK299737.1 | - | 19 - 20 | 2409 | 136 | |
BC016392.1 | - | 16 - 17 | 2190 | 136 | |
BC112387.1 | - | 7 - 8 | 945 | 136 | |
DQ341187.1 | - | 47 - 48 | 6193 | 136 |