The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271999.1 | NP_001258928.1 | 26 - 27 | 3287 | 86 |
NM_001272000.1 | NP_001258929.1 | 25 - 26 | 3194 | 86 | |
NM_001272001.1 | NP_001258930.1 | 25 - 26 | 3194 | 86 | |
NM_033407.3 | NP_212132.2 | 25 - 26 | 3194 | 86 | |
XM_005271292.2 | XP_005271349.1 | 26 - 27 | 3277 | 86 | |
XM_011542326.2 | XP_011540628.1 | 26 - 27 | 3277 | 86 | |
XM_011542327.2 | XP_011540629.1 | 26 - 27 | 3277 | 86 | |
XM_011542328.2 | XP_011540630.1 | 26 - 27 | 3277 | 86 | |
XM_011542330.2 | XP_011540632.1 | 26 - 27 | 3277 | 86 | |
XM_017002639.1 | XP_016858128.1 | 25 - 26 | 3184 | 86 | |
XM_017002640.1 | XP_016858129.1 | 26 - 27 | 3277 | 86 | |
GenBank mRNA | AB051558.1 | - | 6 - 7 | 697 | 86 |
AK125049.1 | - | 11 - 12 | 1339 | 86 | |
DQ118679.1 | - | 22 - 23 | 2766 | 86 | |
DQ118680.1 | - | 22 - 23 | 2766 | 86 | |
DQ341187.1 | - | 26 - 27 | 3194 | 86 |