The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152309.2 | NP_689522.2 | 2 - 3 | 561 | 73 |
XM_011539248.1 | XP_011537550.1 | 2 - 3 | 553 | 73 | |
XM_011539249.1 | XP_011537551.1 | 2 - 3 | 553 | 73 | |
GenBank mRNA | AK125635.1 | - | 2 - 3 | 553 | 73 |
BX648550.1 | - | 2 - 3 | 457 | 73 |