Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIK3AP1
Entrez Gene ID
118788
Gene Name
phosphoinositide-3-kinase adaptor protein 1
Gene Aliases
BCAP
Chromosome Location
Chr.10: 96593312 - 96720522 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152309.2 NP_689522.2 4 - 5 838 100
XM_005269498.1 XP_005269555.1 3 - 4 361 100
XM_005269499.1 XP_005269556.1 3 - 4 385 100
XM_011539248.1 XP_011537550.1 4 - 5 830 100
XM_011539249.1 XP_011537551.1 4 - 5 830 100
GenBank mRNA AK092883.1 - 3 - 4 361 100
AK125635.1 - 4 - 5 830 100
BX648550.1 - 4 - 5 734 100

View Full Product Details