Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATG4B
Entrez Gene ID
23192
Gene Name
autophagy related 4B cysteine peptidase
Gene Aliases
APG4B, AUTL1
Chromosome Location
Chr.2: 241637612 - 241673857 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013325.4 NP_037457.3 6 - 7 557 60
NM_178326.2 NP_847896.1 6 - 7 557 60
XM_005246992.3 XP_005247049.2 6 - 7 543 60
XM_005246993.2 XP_005247050.2 6 - 7 543 60
XM_005246995.3 XP_005247052.1 7 - 8 594 60
XM_005246996.2 XP_005247053.1 7 - 8 539 60
XM_017003637.1 XP_016859126.1 6 - 7 656 60
XM_017003638.1 XP_016859127.1 6 - 7 655 60
XM_017003639.1 XP_016859128.1 7 - 8 613 60
XM_017003640.1 XP_016859129.1 8 - 9 619 60
GenBank mRNA AB023160.2 - 6 - 7 464 60
AB066215.1 - 5 - 6 451 60
AJ504652.1 - 5 - 6 460 60
AK027332.1 - 6 - 7 423 60
AK027462.1 - 6 - 7 470 60
AK027763.1 - 6 - 7 490 60
AK125277.1 - 6 - 7 981 60
AK296818.1 - 7 - 8 539 60
AK298109.1 - 7 - 8 567 60
AK302963.1 - 7 - 8 807 60
AK316167.1 - 3 - 4 315 60
AL080168.1 - 6 - 7 558 60
BC000719.2 - 6 - 7 461 60
BX648007.1 - 7 - 8 538 60

View Full Product Details