The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004714.2 | NP_004705.1 | 4 - 5 | 682 | 67 |
NM_006483.2 | NP_006474.1 | 4 - 5 | 682 | 67 | |
NM_006484.2 | NP_006475.1 | 4 - 5 | 682 | 67 | |
XM_005259398.4 | XP_005259455.1 | 4 - 5 | 1134 | 67 | |
GenBank mRNA | AF205861.1 | - | 4 - 5 | 579 | 67 |
AK074971.1 | - | 4 - 5 | 598 | 67 | |
AK302127.1 | - | 4 - 5 | 611 | 67 | |
BC018751.1 | - | 4 - 5 | 634 | 67 | |
BC025291.1 | - | 4 - 5 | 585 | 67 | |
Y17999.1 | - | 4 - 5 | 650 | 67 |