Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLGN3
Entrez Gene ID
54413
Gene Name
neuroligin 3
Gene Aliases
HNL3
Chromosome Location
Chr.X: 71144389 - 71175307 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181303.1 NP_851820.1 2 - 3 795 66
XM_006724662.3 XP_006724725.2 3 - 4 610 66
XM_006724663.3 XP_006724726.1 3 - 4 802 66
XM_011530974.2 XP_011529276.1 2 - 3 724 66
XM_017029597.1 XP_016885086.1 1 - 2 597 66
GenBank mRNA AF217412.1 - 1 - 2 507 66
GQ489207.1 - 1 - 2 461 66
KJ534891.1 - 1 - 2 461 66

View Full Product Details