Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP13A1
Entrez Gene ID
57130
Gene Name
ATPase 13A1
Gene Aliases
ATP13A, CGI-152
Chromosome Location
Chr.19: 19645198 - 19663694 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020410.2 NP_065143.2 11 - 12 1560 86
GenBank mRNA AB058728.1 - 11 - 12 1530 86
AF288687.1 - 11 - 12 1178 86
AK056420.1 - 11 - 12 1536 86
AK074159.1 - 4 - 5 596 86
AK130125.1 - 2 - 3 200 86

View Full Product Details