The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020410.2 | NP_065143.2 | 13 - 14 | 1840 | 117 |
GenBank mRNA | AB058728.1 | - | 13 - 14 | 1810 | 117 |
AF288687.1 | - | 13 - 14 | 1458 | 117 | |
AK056420.1 | - | 13 - 14 | 1816 | 117 | |
AK074798.1 | - | 2 - 3 | 275 | 117 | |
AK130125.1 | - | 4 - 5 | 480 | 117 | |
AK172778.1 | - | 12 - 13 | 1271 | 117 |