The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020410.2 | NP_065143.2 | 7 - 8 | 1107 | 74 |
GenBank mRNA | AB058728.1 | - | 7 - 8 | 1077 | 74 |
AF288687.1 | - | 7 - 8 | 725 | 74 | |
AK056420.1 | - | 7 - 8 | 1083 | 74 | |
AK074159.1 | - | 1 - 2 | 69 | 74 |