The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020410.2 | NP_065143.2 | 9 - 10 | 1303 | 59 |
GenBank mRNA | AB058728.1 | - | 9 - 10 | 1273 | 59 |
AF288687.1 | - | 9 - 10 | 921 | 59 | |
AK056420.1 | - | 9 - 10 | 1279 | 59 | |
AK074159.1 | - | 3 - 4 | 265 | 59 | |
AK172778.1 | - | 8 - 9 | 734 | 59 |