Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCN10A
Entrez Gene ID
6336
Gene Name
sodium voltage-gated channel alpha subunit 10
Gene Aliases
FEPS2, Nav1.8, PN3, SNS, hPN3
Chromosome Location
Chr.3: 38697110 - 38794010 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293306.2 NP_001280235.2 7 - 8 951 78
NM_001293307.2 NP_001280236.2 7 - 8 951 78
NM_006514.3 NP_006505.3 7 - 8 951 78
XM_005265371.3 XP_005265428.1 7 - 8 960 78
XM_011533993.2 XP_011532295.1 7 - 8 960 78
XM_011533994.2 XP_011532296.1 7 - 8 960 78
GenBank mRNA AF117907.1 - 7 - 8 951 78

View Full Product Details