The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139056.3 | NP_620687.2 | 15 - 16 | 2419 | 88 |
NR_136935.1 | 14 - 15 | 2288 | 88 | ||
GenBank mRNA | AB095949.1 | - | 12 - 13 | 1674 | 88 |
AJ315734.1 | - | 15 - 16 | 2281 | 88 | |
AK127746.1 | - | 15 - 16 | 2419 | 88 |