The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139056.3 | NP_620687.2 | 5 - 6 | 1100 | 120 |
NR_136935.1 | 5 - 6 | 1100 | 120 | ||
GenBank mRNA | AB095949.1 | - | 2 - 3 | 355 | 120 |
AJ315734.1 | - | 5 - 6 | 962 | 120 | |
AK122980.1 | - | 3 - 4 | 725 | 120 | |
DQ266047.1 | - | 5 - 6 | 1100 | 120 |