The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286138.1 | NP_001273067.1 | 1 - 2 | 406 | 76 |
XM_005266801.2 | XP_005266858.1 | 1 - 2 | 167 | 76 | |
XM_005266802.2 | XP_005266859.1 | 1 - 2 | 2275 | 76 | |
GenBank mRNA | AK301365.1 | - | 1 - 2 | 129 | 76 |