The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039619.2 | NP_001034708.1 | 2 - 3 | 536 | 73 |
NM_001282956.1 | NP_001269885.1 | 2 - 3 | 536 | 73 | |
NM_006109.4 | NP_006100.2 | 2 - 3 | 451 | 73 | |
GenBank mRNA | AB451246.1 | - | 2 - 3 | 227 | 73 |
AB451370.1 | - | 2 - 3 | 227 | 73 | |
AF015913.1 | - | 2 - 3 | 227 | 73 | |
AF167572.1 | - | 2 - 3 | 318 | 73 | |
AK075251.1 | - | 2 - 3 | 329 | 73 | |
AK315500.1 | - | 2 - 3 | 267 | 73 | |
AL833612.1 | - | 2 - 3 | 236 | 73 | |
BC025979.2 | - | 2 - 3 | 239 | 73 | |
CR456741.1 | - | 2 - 3 | 227 | 73 |