The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039619.2 | NP_001034708.1 | 6 - 7 | 923 | 96 |
NM_001282953.1 | NP_001269882.1 | 5 - 6 | 791 | 96 | |
NM_001282954.1 | NP_001269883.1 | 5 - 6 | 804 | 96 | |
NM_001282955.1 | NP_001269884.1 | 5 - 6 | 706 | 96 | |
NM_006109.4 | NP_006100.2 | 6 - 7 | 838 | 96 | |
XM_017020927.1 | XP_016876416.1 | 5 - 6 | 562 | 96 | |
GenBank mRNA | AB451246.1 | - | 6 - 7 | 614 | 96 |
AB451370.1 | - | 6 - 7 | 614 | 96 | |
AF015913.1 | - | 6 - 7 | 614 | 96 | |
AF167572.1 | - | 6 - 7 | 705 | 96 | |
AK075251.1 | - | 6 - 7 | 716 | 96 | |
AK300863.1 | - | 5 - 6 | 588 | 96 | |
AK301812.1 | - | 5 - 6 | 597 | 96 | |
AK302240.1 | - | 5 - 6 | 486 | 96 | |
AK315500.1 | - | 6 - 7 | 654 | 96 | |
AL833612.1 | - | 6 - 7 | 623 | 96 | |
BC025979.2 | - | 6 - 7 | 626 | 96 | |
CR456741.1 | - | 6 - 7 | 614 | 96 |