Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDRG2
Entrez Gene ID
57447
Gene Name
NDRG family member 2
Gene Aliases
SYLD
Chromosome Location
Chr.14: 21016763 - 21070872 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282212.1 NP_001269141.1 1 - 2 247 85
XM_011536996.2 XP_011535298.1 1 - 2 82 85
XM_017021482.1 XP_016876971.1 1 - 2 82 85
GenBank mRNA BX247987.1 - 1 - 2 106 85
DA065447.1 - 1 - 2 247 85

View Full Product Details