The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127397.2 | NP_001120869.1 | 10 - 11 | 1380 | 102 |
NM_001127398.2 | NP_001120870.1 | 9 - 10 | 1218 | 102 | |
NM_015701.4 | NP_056516.2 | 10 - 11 | 1380 | 102 | |
XM_011532766.1 | XP_011531068.1 | 10 - 11 | 1294 | 102 | |
GenBank mRNA | AF131743.1 | - | 2 - 3 | 199 | 102 |
AF131849.1 | - | 4 - 5 | 569 | 102 | |
AK001913.1 | - | 10 - 11 | 1261 | 102 | |
AK315477.1 | - | 10 - 11 | 1169 | 102 | |
AY358717.1 | - | 9 - 10 | 1102 | 102 | |
AY453410.1 | - | 10 - 11 | 1231 | 102 | |
BC013129.1 | - | 10 - 11 | 1231 | 102 | |
BC022228.1 | - | 9 - 10 | 1066 | 102 | |
BQ017438.1 | - | 3 - 4 | 102 | ||
EU794676.1 | - | 10 - 11 | 1380 | 102 |