Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERLEC1
Entrez Gene ID
27248
Gene Name
endoplasmic reticulum lectin 1
Gene Aliases
C2orf30, CIM, CL24936, CL25084, HEL117, XTP3-B, XTP3TPB
Chromosome Location
Chr.2: 53786931 - 53826928 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127397.2 NP_001120869.1 2 - 3 545 75
NM_001127398.2 NP_001120870.1 2 - 3 545 75
NM_015701.4 NP_056516.2 2 - 3 545 75
XM_011532766.1 XP_011531068.1 2 - 3 459 75
GenBank mRNA AK001913.1 - 2 - 3 426 75
AK315477.1 - 2 - 3 334 75
AY358717.1 - 2 - 3 429 75
AY453410.1 - 2 - 3 396 75
BC013129.1 - 2 - 3 396 75
BC022228.1 - 2 - 3 393 75
EU794676.1 - 2 - 3 545 75

View Full Product Details