Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DYRK1B
Entrez Gene ID
9149
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 1B
Gene Aliases
AOMS3, MIRK
Chromosome Location
Chr.19: 39825347 - 39834233 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006484.2 NP_006475.1 9 - 10 1449 87

View Full Product Details