Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRYBA1
Entrez Gene ID
1411
Gene Name
crystallin beta A1
Gene Aliases
CRYB1, CTRCT10
Chromosome Location
Chr.17: 29246857 - 29254494 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005208.4 NP_005199.2 2 - 3 104 93
XM_017024198.1 XP_016879687.1 3 - 4 400 93
GenBank mRNA BC069537.1 - 2 - 3 97 93
BC113693.1 - 2 - 3 98 93
KM230602.1 - 2 - 3 86 93
KM230603.1 - 2 - 3 85 93
KM230604.1 - 2 - 3 82 93
KM230605.1 - 1 - 2 82 93
KR336660.1 - 2 - 3 82 93

View Full Product Details