Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRYBA1
Entrez Gene ID
1411
Gene Name
crystallin beta A1
Gene Aliases
CRYB1, CTRCT10
Chromosome Location
Chr.17: 29246857 - 29254494 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005208.4 NP_005199.2 5 - 6 504 101
XM_017024198.1 XP_016879687.1 6 - 7 800 101
GenBank mRNA BC069537.1 - 5 - 6 497 101
BC113693.1 - 5 - 6 498 101
KM230602.1 - 5 - 6 486 101
KM230603.1 - 5 - 6 485 101
KM230604.1 - 5 - 6 482 101
KM230605.1 - 4 - 5 482 101
KR336660.1 - 5 - 6 482 101
KR336661.1 - 2 - 3 171 101
U59058.1 - 3 - 4 370 101

View Full Product Details