The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006277.2 | NP_006268.2 | 25 - 26 | 3374 | 163 |
NM_019595.3 | NP_062541.3 | 24 - 25 | 3293 | 163 | |
NM_147152.2 | NP_671494.2 | 25 - 26 | 3374 | 163 | |
GenBank mRNA | AB033082.1 | - | 24 - 25 | 3200 | 163 |
AF001630.1 | - | 5 - 6 | 653 | 163 | |
AF038189.1 | - | 5 - 6 | 776 | 163 | |
AF182198.1 | - | 25 - 26 | 3353 | 163 | |
AF182199.1 | - | 25 - 26 | 3353 | 163 | |
AF248540.1 | - | 23 - 24 | 3074 | 163 | |
BC146779.1 | - | 24 - 25 | 3200 | 163 | |
U61167.1 | - | 3 - 4 | 284 | 163 |