The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000918.3 | NP_000909.2 | 6 - 7 | 1049 | 57 |
GenBank mRNA | AK075503.1 | - | - | 1454 | 57 |
AK095938.1 | - | 6 - 7 | 927 | 57 | |
AK128783.1 | - | 3 - 4 | 297 | 57 | |
AK297081.1 | - | 6 - 7 | 775 | 57 | |
AK297967.1 | - | 5 - 6 | 869 | 57 | |
AK298005.1 | - | 5 - 6 | 805 | 57 | |
AK300561.1 | - | 5 - 6 | 779 | 57 | |
AK308600.1 | - | 4 - 5 | 704 | 57 | |
AK315631.1 | - | 6 - 7 | 880 | 57 | |
BC010859.1 | - | 6 - 7 | 896 | 57 | |
BC014504.1 | - | 2 - 3 | 148 | 57 | |
BC029617.1 | - | 6 - 7 | 896 | 57 | |
BC071892.1 | - | 6 - 7 | 855 | 57 | |
J02783.1 | - | 6 - 7 | 915 | 57 | |
X05130.1 | - | 6 - 7 | 881 | 57 |