Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFR
Entrez Gene ID
51663
Gene Name
zinc finger RNA binding protein
Gene Aliases
SPG71, ZFR1
Chromosome Location
Chr.5: 32354350 - 32444738 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016107.3 NP_057191.2 13 - 14 2419 78
GenBank mRNA AF100742.1 - 3 - 4 414 78
AJ314790.1 - 13 - 14 2372 78
AK025481.1 - 1 - 2 112 78
AK295297.1 - 1 - 2 63 78
BC051893.1 - 6 - 7 1010 78
BC062986.1 - 6 - 7 935 78
BC137084.1 - 13 - 14 2383 78
BC137085.1 - 13 - 14 2383 78

View Full Product Details