Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFR
Entrez Gene ID
51663
Gene Name
zinc finger RNA binding protein
Gene Aliases
SPG71, ZFR1
Chromosome Location
Chr.5: 32354350 - 32444738 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016107.3 NP_057191.2 14 - 15 2580 83
GenBank mRNA AJ314790.1 - 14 - 15 2533 83
AK025481.1 - 2 - 3 273 83
AK295297.1 - 2 - 3 224 83
BC051893.1 - 7 - 8 1171 83
BC062986.1 - 7 - 8 1096 83
BC137084.1 - 14 - 15 2544 83
BC137085.1 - 14 - 15 2544 83

View Full Product Details