Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZFR
Entrez Gene ID
51663
Gene Name
zinc finger RNA binding protein
Gene Aliases
SPG71, ZFR1
Chromosome Location
Chr.5: 32354350 - 32444738 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016107.3 NP_057191.2 1 - 2 121 72
GenBank mRNA AJ314790.1 - 1 - 2 74 72
AK292214.1 - 1 - 2 121 72
AK309638.1 - 1 - 2 121 72
BC017879.1 - 1 - 2 121 72
BC030540.1 - 1 - 2 93 72
BC137084.1 - 1 - 2 85 72
BC137085.1 - 1 - 2 85 72

View Full Product Details