Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM40
Entrez Gene ID
135644
Gene Name
tripartite motif containing 40
Gene Aliases
RNF35
Chromosome Location
Chr.6: 30135998 - 30148773 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011514306.1 XP_011512608.1 2 - 3 378 90

View Full Product Details