Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM40
Entrez Gene ID
135644
Gene Name
tripartite motif containing 40
Gene Aliases
RNF35
Chromosome Location
Chr.6: 30135998 - 30148773 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286633.1 NP_001273562.1 5 - 6 1076 132
NM_138700.4 NP_619645.1 4 - 5 907 132
XM_011514305.1 XP_011512607.1 5 - 6 1507 132
XM_011514306.1 XP_011512608.1 6 - 7 1373 132
GenBank mRNA AF489517.1 - 4 - 5 630 188
BC060785.1 - 5 - 6 1076 132

View Full Product Details