The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013694.2 | NP_001013716.2 | 1 - 2 | 216 | 110 |
XM_011530178.2 | XP_011528480.1 | 1 - 2 | 233 | 156 | |
XM_017028799.1 | XP_016884288.1 | 1 - 2 | 228 | 110 | |
GenBank mRNA | AK293916.1 | - | 2 - 3 | 196 | 110 |
DA325985.1 | - | 1 - 2 | 211 | 110 |