Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TEAD2
- Entrez Gene ID
-
8463
- Gene Name
- TEA domain transcription factor 2
- Gene Aliases
-
ETF, TEAD-2, TEF-4, TEF4
- Chromosome Location
-
Chr.19: 49340595 - 49362457 on Build GRCh38
View Full Product Details